accessibility menu, dialog, popup

UserName

CAS RN 17635-44-8

IUPAC Name: 3,4,5-tribromo-1H-pyrazole
Synonyms: 3,4,5-tribromo-1H-pyrazole
Molecular Weight (g/mol): 304.77
Molecular Formula: C3HBr3N2
InChi Key: TXQKCKQJBGFUBF-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(Br)=NN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)