missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Tribromopyrazole 98.0+%, TCI America™

Catalog No. p-7137410
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27775G 5 g
T277725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27775G Supplier TCI America Supplier No. T27775G
Only null left

Chemical Identifiers

CAS 17635-44-8
Molecular Formula C3HBr3N2
Molecular Weight (g/mol) 304.767
MDL Number MFCD00040248
InChI Key TXQKCKQJBGFUBF-UHFFFAOYSA-N
Synonym 3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g
PubChem CID 627674
IUPAC Name 3,4,5-tribromo-1H-pyrazole
SMILES C1(=C(NN=C1Br)Br)Br

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 5 g
Formula Weight 304.77
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Tribromopyrazole