missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Thiodipropionic Acid 99.0+%, TCI America™
Supplier: TCI America T0204500G
Specifications
| 3,3′-Thiodipropionic Acid | |
| 131°C | |
| C6H10O4S | |
| 500 g | |
| ODJQKYXPKWQWNK-UHFFFAOYSA-N | |
| 3-(2-carboxyethylsulfanyl)propanoic acid | |
| 8096 | |
| ≥99.0% (T) |
| 111-17-1 | |
| White | |
| MFCD00002781 | |
| 3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid | |
| C(CSCCC(=O)O)C(=O)O | |
| 178.202 | |
| 178.20 | |
| Crystalline Powder |
Chemical Identifiers
| 111-17-1 | |
| 178.202 | |
| ODJQKYXPKWQWNK-UHFFFAOYSA-N | |
| 8096 | |
| C(CSCCC(=O)O)C(=O)O |
| C6H10O4S | |
| MFCD00002781 | |
| 3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid | |
| 3-(2-carboxyethylsulfanyl)propanoic acid |
Safety and Handling
EINECSNumber : (2)-0920
RTECSNumber : UF7990000
TSCA : Yes