missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Thiodipropionic Acid 99.0+%, TCI America™

Catalog No. p-7136143
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T020425G 25 g
T0204500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T020425G Supplier TCI America Supplier No. T020425G
Only null left

Chemical Identifiers

CAS 111-17-1
Molecular Formula C6H10O4S
Molecular Weight (g/mol) 178.202
MDL Number MFCD00002781
InChI Key ODJQKYXPKWQWNK-UHFFFAOYSA-N
Synonym 3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
PubChem CID 8096
IUPAC Name 3-(2-carboxyethylsulfanyl)propanoic acid
SMILES C(CSCCC(=O)O)C(=O)O

Specifications

Melting Point 131°C
Color White
Quantity 25 g
Formula Weight 178.20
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Thiodipropionic Acid
EINECSNumber (2)-0920
RTECSNumber UF7990000
TSCA Yes