missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3-Nitrophenyl)propionic Acid 98.0+%, TCI America™
Supplier: TCI America N096025G
Specifications
| 3-(3-Nitrophenyl)propionic Acid | |
| 119°C | |
| C9H9NO4 | |
| 25 g | |
| ZOANOABZUNJOJT-UHFFFAOYSA-N | |
| 195.174 | |
| 195.17 | |
| Crystalline Powder |
| 1664-57-9 | |
| White-Yellow | |
| MFCD01310835 | |
| 3-Nitrohydrocinnamic Acid | |
| C1=CC(=CC(=C1)[N+](=O)[O-])CCC(=O)O | |
| 2760224 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 1664-57-9 | |
| 195.174 | |
| ZOANOABZUNJOJT-UHFFFAOYSA-N | |
| 2760224 |
| C9H9NO4 | |
| MFCD01310835 | |
| 3-Nitrohydrocinnamic Acid | |
| C1=CC(=CC(=C1)[N+](=O)[O-])CCC(=O)O |
Safety and Handling
TSCA : No