missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Nitrophenyl)propionic Acid 98.0+%, TCI America™

Catalog No. p-7134050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09605G 5 g
N096025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09605G Supplier TCI America Supplier No. N09605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1664-57-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD01310835
InChI Key ZOANOABZUNJOJT-UHFFFAOYSA-N
Synonym 3-Nitrohydrocinnamic Acid
PubChem CID 2760224
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CCC(=O)O

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(3-Nitrophenyl)propionic Acid