accessibility menu, dialog, popup

UserName

NOOOHO

CAS RN 1664-57-9

IUPAC Name: 3-(3-nitrophenyl)propanoic acid
Synonyms: 3-(3-nitrophenyl)propanoic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: ZOANOABZUNJOJT-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(=CC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results