missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenylpropionic Acid 97.0+%, TCI America™

Catalog No. D177925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D177925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D177925G Supplier TCI America Supplier No. D177925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 606-83-7
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00002717
InChI Key BZQGAPWJKAYCHR-UHFFFAOYSA-N
Synonym 3,3-diphenylpropionic acid, benzenepropanoic acid, .beta.-phenyl, 3,3-diphenyl-propionic acid, propionic acid, 3,3-diphenyl, .beta.-phenylbenzenepropanoic acid, beta,beta-diphenylpropionic acid, hydrocinnamic acid, .beta.-phenyl, beta-phenylbenzenepropanoic acid, hydrocinnamic acid, beta-phenyl, pubchem13618
PubChem CID 64798
IUPAC Name 3,3-diphenylpropanoic acid
SMILES OC(=O)CC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 153°C
Color White
Quantity 25 g
Formula Weight 226.28
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Diphenylpropionic Acid