accessibility menu, dialog, popup

UserName

HOO

CAS RN 606-83-7

IUPAC Name: 3,3-diphenylpropanoic acid
Synonyms: 3,3-diphenylpropanoic acid
Molecular Weight (g/mol): 226.28
Molecular Formula: C15H14O2
InChi Key: BZQGAPWJKAYCHR-UHFFFAOYSA-N
SMILES: OC(=O)CC(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)