missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Diphenyl-1-propanol 98.0+%, TCI America™
$516.35 - $1668.33
Chemical Identifiers
| CAS | 20017-67-8 |
|---|---|
| Molecular Formula | C15H16O |
| Molecular Weight (g/mol) | 212.29 |
| MDL Number | MFCD00002930 |
| InChI Key | IDCXQMVSIIJUEH-UHFFFAOYSA-N |
| PubChem CID | 29908 |
| IUPAC Name | 3,3-diphenylpropan-1-ol |
| SMILES | OCCC(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 20017-67-8 | |
| 212.29 | |
| IDCXQMVSIIJUEH-UHFFFAOYSA-N | |
| 3,3-diphenylpropan-1-ol |
| C15H16O | |
| MFCD00002930 | |
| 29908 | |
| OCCC(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 20017-67-8 | |
| 208°C | |
| MFCD00002930 | |
| IDCXQMVSIIJUEH-UHFFFAOYSA-N | |
| 3,3-diphenylpropan-1-ol | |
| 29908 | |
| ≥98.0% (GC) | |
| 3,3-Diphenyl-1-propanol |
| Yellow | |
| C15H16O | |
| 5 g | |
| OCCC(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 212.29 | |
| 212.29 | |
| Liquid |
Safety and Handling
TSCA : No