accessibility menu, dialog, popup

UserName

CAS RN 20017-67-8

HO

CAS RN 20017-67-8

IUPAC Name: 3,3-diphenylpropan-1-ol
Synonyms: 3,3-diphenylpropan-1-ol
Molecular Weight (g/mol): 212.29
Molecular Formula: C15H16O
InChi Key: IDCXQMVSIIJUEH-UHFFFAOYSA-N
SMILES: OCCC(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results