missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dinitrobenzophenone 99.0+%, TCI America™

Catalog No. D16885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16885G 5 g
D168825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16885G Supplier TCI America Supplier No. D16885G

Chemical Identifiers

CAS 21222-05-9
Molecular Formula C13H8N2O5
Molecular Weight (g/mol) 272.22
MDL Number MFCD00039740
InChI Key BSDKBWGNIJMCID-UHFFFAOYSA-N
Synonym 3,3'-dinitrobenzophenone, bis 3-nitrophenyl methanone, methanone, bis 3-nitrophenyl, methanone, bis 3-nitrophenyl-9ci, di3-nitrophenyl ketone, 3,3-dinitrobenzophenone, 3,3/'-dinitrobenzophenone, acmc-1cp75, bis 3-nitrophenyl methanone #, methanone,bis 3-nitrophenyl
PubChem CID 96690
IUPAC Name bis(3-nitrophenyl)methanone
SMILES [O-][N+](=O)C1=CC(=CC=C1)C(=O)C1=CC(=CC=C1)[N+]([O-])=O

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 5 g
Formula Weight 272.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dinitrobenzophenone