CAS RN 21222-05-9
IUPAC Name:
bis(3-nitrophenyl)methanone
Synonyms:
bis(3-nitrophenyl)methanone
Molecular Weight (g/mol):
272.22
Molecular Formula:
C13H8N2O5
InChi Key:
BSDKBWGNIJMCID-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(=CC=C1)C(=O)C1=CC(=CC=C1)[N+]([O-])=O
1
–
1
of
1
results
1
3,3'-Dinitrobenzophenone 99.0+%, TCI America™
CAS: 21222-05-9 Molecular Formula: C13H8N2O5 Molecular Weight (g/mol): 272.22 MDL Number: MFCD00039740 InChI Key: BSDKBWGNIJMCID-UHFFFAOYSA-N Synonym: 3,3'-dinitrobenzophenone,bis 3-nitrophenyl methanone,methanone, bis 3-nitrophenyl,methanone, bis 3-nitrophenyl-9ci,di3-nitrophenyl ketone,3,3-dinitrobenzophenone,3,3/'-dinitrobenzophenone,acmc-1cp75,bis 3-nitrophenyl methanone #,methanone,bis 3-nitrophenyl PubChem CID: 96690 IUPAC Name: bis(3-nitrophenyl)methanone SMILES: [O-][N+](=O)C1=CC(=CC=C1)C(=O)C1=CC(=CC=C1)[N+]([O-])=O