missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Dimethylamino)propoxy]benzylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC59413DA

 View more versions of this product

Catalog No. CC59413DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-[3-(Dimethylamino)propoxy]benzylamine
94.5
95%
C12H20N2O
1g
VRMIFLYRNDVHIN-UHFFFAOYSA-N
1-{3-[3-(dimethylamino)propoxy]phenyl}methanamine
21465371
95%
182963-94-6
100.0
Amber Glass Bottle
MFCD09064990
3-3-dimethylamino propoxy benzylamine, 3-3-aminomethyl phenoxy-n,n-dimethylpropan-1-amine, 3-3-dimethylamino propoxy phenyl methanamine, 3-3-aminomethyl phenoxy propyl dimethylamine, benzenemethanamine,3-3-dimethylamino propoxy, 3-3-aminomethyl phenoxy-n,n-dimethyl-propan-1-amine
CN(C)CCCOC1=CC=CC(CN)=C1
208.31
208.3

Chemical Identifiers

182963-94-6
208.31
VRMIFLYRNDVHIN-UHFFFAOYSA-N
21465371
CN(C)CCCOC1=CC=CC(CN)=C1
C12H20N2O
MFCD09064990
3-3-dimethylamino propoxy benzylamine, 3-3-aminomethyl phenoxy-n,n-dimethylpropan-1-amine, 3-3-dimethylamino propoxy phenyl methanamine, 3-3-aminomethyl phenoxy propyl dimethylamine, benzenemethanamine,3-3-dimethylamino propoxy, 3-3-aminomethyl phenoxy-n,n-dimethyl-propan-1-amine
1-{3-[3-(dimethylamino)propoxy]phenyl}methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions