missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Dimethylamino)propoxy]benzylamine, 97%, Thermo Scientific™

Catalog No. p-3522485
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC59413CB 250mg Amber Glass Bottle
CC59413DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC59413CB Supplier Thermo Scientific Chemicals Supplier No. CC59413CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 182963-94-6
Molecular Formula C12H20N2O
Molecular Weight (g/mol) 208.31
MDL Number MFCD09064990
InChI Key VRMIFLYRNDVHIN-UHFFFAOYSA-N
Synonym 3-3-dimethylamino propoxy benzylamine, 3-3-aminomethyl phenoxy-n,n-dimethylpropan-1-amine, 3-3-dimethylamino propoxy phenyl methanamine, 3-3-aminomethyl phenoxy propyl dimethylamine, benzenemethanamine,3-3-dimethylamino propoxy, 3-3-aminomethyl phenoxy-n,n-dimethyl-propan-1-amine
PubChem CID 21465371
IUPAC Name 3-[3-(aminomethyl)phenoxy]-N,N-dimethylpropan-1-amine
SMILES CN(C)CCCOC1=CC=CC(CN)=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 208.3
Percent Purity 95%
Chemical Name or Material 3-[3-(Dimethylamino)propoxy]benzylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.