CAS RN 182963-94-6
IUPAC Name:
1-{3-[3-(dimethylamino)propoxy]phenyl}methanamine
Synonyms:
1-{3-[3-(dimethylamino)propoxy]phenyl}methanamine
Molecular Weight (g/mol):
208.31
Molecular Formula:
C12H20N2O
InChi Key:
VRMIFLYRNDVHIN-UHFFFAOYSA-N
SMILES:
CN(C)CCCOC1=CC=CC(CN)=C1
1
–
1
of
1
results
1
3-[3-(Dimethylamino)propoxy]benzylamine, 97%, Thermo Scientific™
CAS: 182963-94-6 Molecular Formula: C12H20N2O Molecular Weight (g/mol): 208.31 MDL Number: MFCD09064990 InChI Key: VRMIFLYRNDVHIN-UHFFFAOYSA-N Synonym: 3-3-dimethylamino propoxy benzylamine,3-3-aminomethyl phenoxy-n,n-dimethylpropan-1-amine,3-3-dimethylamino propoxy phenyl methanamine,3-3-aminomethyl phenoxy propyl dimethylamine,benzenemethanamine,3-3-dimethylamino propoxy,3-3-aminomethyl phenoxy-n,n-dimethyl-propan-1-amine PubChem CID: 21465371 IUPAC Name: 1-{3-[3-(dimethylamino)propoxy]phenyl}methanamine SMILES: CN(C)CCCOC1=CC=CC(CN)=C1