missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethoxybenzidine, 97%

Catalog No. p-114488
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407890250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407890250 Supplier Thermo Scientific Chemicals Supplier No. 407890250
Only null left

CAS: 119-90-4 | C14H16N2O2 | 244.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-90-4
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.29
MDL Number MFCD00008372
InChI Key JRBJSXQPQWSCCF-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzidine, o-dianisidine, dianisidine, cellitazol b, neutrosel navy bn, diacel navy dc, disperse black 6, blue base nb, blue bn base, blue base irga b
PubChem CID 8411
ChEBI CHEBI:82321
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 135°C to 139°C
Color Beige-Brown to Pink
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 2992
Formula Weight 244.29
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3'-Dimethoxybenzidine, pract., 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause cancer.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-355-4
RTECSNumber DD0875000
TSCA TSCA
Recommended Storage Product darkens on storage

RUO – Research Use Only