missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dihydroxybenzidine 99.0+%, TCI America™

Catalog No. p-7127223
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23125G 5 g
D231225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23125G Supplier TCI America Supplier No. D23125G
Only null left

Chemical Identifiers

CAS 2373-98-0
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD00039149
InChI Key ZGDMDBHLKNQPSD-UHFFFAOYSA-N
Synonym Benzidine-3,3′C-diol, 4,4′C-Diamino-3,3′C-dihydroxybiphenyl
PubChem CID 16918
IUPAC Name 4,4'-diamino-[1,1'-biphenyl]-3,3'-diol
SMILES NC1=CC=C(C=C1O)C1=CC=C(N)C(O)=C1

Specifications

Melting Point 292°C
Quantity 5 g
Formula Weight 216.24
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dihydroxybenzidine
RTECSNumber DV4900000
TSCA No