CAS RN 2373-98-0
CAS RN 2373-98-0
IUPAC Name:
4,4'-diamino-[1,1'-biphenyl]-3,3'-diol
Synonyms:
3,3'-dihydroxybenzidine
Molecular Weight (g/mol):
216.24
Molecular Formula:
C12H12N2O2
InChi Key:
ZGDMDBHLKNQPSD-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1O)C1=CC=C(N)C(O)=C1
1
–
1
of
1
results
1
3,3'-Dihydroxybenzidine 99.0+%, TCI America™
CAS: 2373-98-0 Molecular Formula: C12H12N2O2 Molecular Weight (g/mol): 216.24 MDL Number: MFCD00039149 InChI Key: ZGDMDBHLKNQPSD-UHFFFAOYSA-N Synonym: Benzidine-3,3′C-diol, 4,4′C-Diamino-3,3′C-dihydroxybiphenyl PubChem CID: 16918 IUPAC Name: 4,4'-diamino-[1,1'-biphenyl]-3,3'-diol SMILES: NC1=CC=C(C=C1O)C1=CC=C(N)C(O)=C1