missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Dibromo-4,4'-dimethoxybiphenyl 95.0+%, TCI America™
$202.15 - $646.85
Chemical Identifiers
| CAS | 108989-36-2 |
|---|---|
| Molecular Formula | C14H12Br2O2 |
| Molecular Weight (g/mol) | 372.056 |
| InChI Key | VUOOJGFHEDZGEA-UHFFFAOYSA-N |
| PubChem CID | 15040359 |
| IUPAC Name | 2-bromo-4-(3-bromo-4-methoxyphenyl)-1-methoxybenzene |
| SMILES | COC1=C(C=C(C=C1)C2=CC(=C(C=C2)OC)Br)Br |
Chemical Identifiers
| 108989-36-2 | |
| 372.056 | |
| 15040359 | |
| COC1=C(C=C(C=C1)C2=CC(=C(C=C2)OC)Br)Br |
| C14H12Br2O2 | |
| VUOOJGFHEDZGEA-UHFFFAOYSA-N | |
| 2-bromo-4-(3-bromo-4-methoxyphenyl)-1-methoxybenzene |
Specifications
| 108989-36-2 | |
| White-Yellow | |
| 1 g | |
| COC1=C(C=C(C=C1)C2=CC(=C(C=C2)OC)Br)Br | |
| 372.056 | |
| 372.06 | |
| Crystalline Powder |
| 166°C | |
| C14H12Br2O2 | |
| VUOOJGFHEDZGEA-UHFFFAOYSA-N | |
| 2-bromo-4-(3-bromo-4-methoxyphenyl)-1-methoxybenzene | |
| 15040359 | |
| ≥95.0% (GC) | |
| 3,3′-Dibromo-4,4′-dimethoxybiphenyl |
Safety and Handling
TSCA : Yes