accessibility menu, dialog, popup

UserName

H3COOCH3BrBr

CAS RN 108989-36-2

IUPAC Name: 3,3'-dibromo-4,4'-dimethoxy-1,1'-biphenyl
Synonyms: 3,3'-dibromo-4,4'-dimethoxy-1,1'-biphenyl
Molecular Weight (g/mol): 372.06
Molecular Formula: C14H12Br2O2
InChi Key: VUOOJGFHEDZGEA-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(C=C1)C1=CC(Br)=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results