missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Dibromo-4,4'-dimethoxybiphenyl 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 166°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 372.06 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,3′-Dibromo-4,4′-dimethoxybiphenyl |
Safety and Handling
| TSCA | Yes |