missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3″-Dibromo-1,1':2',1″-terphenyl 98.0+%, TCI America™
Supplier: TCI America D4997200MG
Specifications
| 3,3′′-Dibromo-1,1′:2′,1′′-terphenyl | |
| 92°C | |
| C18H12Br2 | |
| 200 mg | |
| 3,3′C′C-Dibromo-o-terphenyl | |
| C1=CC=C(C(=C1)C2=CC(=CC=C2)Br)C3=CC(=CC=C3)Br | |
| 388.102 | |
| 388.10 | |
| Crystalline Powder |
| 95918-90-4 | |
| White-Yellow | |
| MFCD28138090 | |
| 3152 | |
| GZOLRAQBGRLGHQ-UHFFFAOYSA-N | |
| 1,2-bis(3-bromophenyl)benzene | |
| 58627207 | |
| ≥98.0% (GC) |
Safety and Handling
TSCA : No