CAS RN 95918-90-4
CAS RN 95918-90-4
IUPAC Name:
3'-bromo-2-(3-bromophenyl)-1,1'-biphenyl
Synonyms:
3'-bromo-2-(3-bromophenyl)-1,1'-biphenyl
Molecular Weight (g/mol):
388.1
Molecular Formula:
C18H12Br2
InChi Key:
GZOLRAQBGRLGHQ-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=C1)C1=CC=CC=C1C1=CC(Br)=CC=C1