missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Diaminodiphenyl sulfone, 98%
Description
3,3'-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,3′-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
3,3′-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 171°C to 174°C |
| Odor | Odorless |
| Quantity | 100 g |
| Beilstein | 2215938 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 248.3 |
| Percent Purity | 98% |
| Chemical Name or Material | 3,3'-Diaminodiphenyl sulfone |
Safety and Handling
| Hazard Category | H302-H371-H373 |
| Precautionary Statement | P260-P264b-P270-P301+P312-P309+P311-P330-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 209-967-5 |
| RTECSNumber | BY8800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only