missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminodiphenyl sulfone, 98%

Catalog No. AAL1140014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1140014 25 g
AAL1140022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1140014 Supplier Thermo Scientific Chemicals Supplier No. L1140014

CAS: 599-61-1 | C12H12N2O2S | 248.30 g/mol

3,3'-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 599-61-1
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.30
MDL Number MFCD00007792
InChI Key LJGHYPLBDBRCRZ-UHFFFAOYSA-N
Synonym 3,3'-sulfonyldianiline, 3,3'-diaminodiphenyl sulfone, 3-aminophenyl sulfone, benzenamine, 3,3'-sulfonylbis, 3,3'-sulphonyldianiline, bis m-aminophenyl sulfone, 3-aminophenyl-sulfone, 3,3'-diaminophenyl sulfone, 3,3'-sulfonylbis aniline, sulfone, bis m-aminophenyl
PubChem CID 11741
IUPAC Name 3-(3-aminophenyl)sulfonylaniline
SMILES NC1=CC(=CC=C1)S(=O)(=O)C1=CC(N)=CC=C1

Specifications

Melting Point 171°C to 174°C
Odor Odorless
Quantity 25 g
Beilstein 2215938
Solubility Information Insoluble in water.
Formula Weight 248.3
Percent Purity 98%
Chemical Name or Material 3,3'-Diaminodiphenyl sulfone
Hazard Category H302-H371-H373
Precautionary Statement P260-P264b-P270-P301+P312-P309+P311-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 209-967-5
RTECSNumber BY8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only