missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzophenone 95.0+%, TCI America™

Catalog No. D16821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16821G 1 g
D16825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16821G Supplier TCI America Supplier No. D16821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-79-0
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00014774
InChI Key TUQQUUXMCKXGDI-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzophenone, bis 3-aminophenyl methanone, methanone, bis 3-aminophenyl, di3-aminophenyl ketone, 3,3'-diaminodiphenyl ketone, pubchem3381, acmc-1awhc, cbmicro_014188, 3,3'-diamino benzophenone, 3-3-aminobenzoyl aniline
PubChem CID 69145
IUPAC Name 3-(3-aminobenzoyl)aniline
SMILES NC1=CC(=CC=C1)C(=O)C1=CC(N)=CC=C1

Specifications

Melting Point 153°C
Color Yellow
Quantity 1 g
Formula Weight 212.25
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diaminobenzophenone
EINECSNumber (4)-1639
TSCA No