accessibility menu, dialog, popup

UserName

NH2ONH2

CAS RN 611-79-0

IUPAC Name: 3-(3-aminobenzoyl)aniline
Synonyms: 3,3'-diaminodiphenyl ketone
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: TUQQUUXMCKXGDI-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1)C(=O)C1=CC(N)=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)