missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzophenone, 90%

CAS: 611-79-0 | C13H12N2O | 212.25 g/mol

Supplier:  Thermo Scientific Chemicals L0912806

 View more versions of this product

Catalog No. AAL0912806


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,3'-Diaminobenzophenone
148°C to 152°C
C13H12N2O
5 g
3,3'-diaminobenzophenone, bis 3-aminophenyl methanone, methanone, bis 3-aminophenyl, di3-aminophenyl ketone, 3,3'-diaminodiphenyl ketone, pubchem3381, acmc-1awhc, cbmicro_014188, 3,3'-diamino benzophenone, 3-3-aminobenzoyl aniline
NC1=CC(=CC=C1)C(=O)C1=CC(N)=CC=C1
212.25
212.25
611-79-0
285°C (11 mmHg)
MFCD00014774
2837605
TUQQUUXMCKXGDI-UHFFFAOYSA-N
bis(3-aminophenyl)methanone
69145
≥90%

Chemical Identifiers

611-79-0
212.25
TUQQUUXMCKXGDI-UHFFFAOYSA-N
69145
NC1=CC(=CC=C1)C(=O)C1=CC(N)=CC=C1
C13H12N2O
MFCD00014774
3,3'-diaminobenzophenone, bis 3-aminophenyl methanone, methanone, bis 3-aminophenyl, di3-aminophenyl ketone, 3,3'-diaminodiphenyl ketone, pubchem3381, acmc-1awhc, cbmicro_014188, 3,3'-diamino benzophenone, 3-3-aminobenzoyl aniline
bis(3-aminophenyl)methanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-281-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only