missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzophenone, 90%

Catalog No. AAL0912803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0912803 1 g
AAL0912806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0912803 Supplier Thermo Scientific Chemicals Supplier No. L0912803

CAS: 611-79-0 | C13H12N2O | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-79-0
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00014774
InChI Key TUQQUUXMCKXGDI-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzophenone, bis 3-aminophenyl methanone, methanone, bis 3-aminophenyl, di3-aminophenyl ketone, 3,3'-diaminodiphenyl ketone, pubchem3381, acmc-1awhc, cbmicro_014188, 3,3'-diamino benzophenone, 3-3-aminobenzoyl aniline
PubChem CID 69145
IUPAC Name bis(3-aminophenyl)methanone
SMILES NC1=CC(=CC=C1)C(=O)C1=CC(N)=CC=C1

Specifications

Melting Point 148°C to 152°C
Boiling Point 285°C (11 mmHg)
Quantity 1 g
Beilstein 2837605
Formula Weight 212.25
Percent Purity ≥90%
Chemical Name or Material 3,3'-Diaminobenzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-281-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only