missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine 98.0+%, TCI America™

Catalog No. p-7136556
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10231G 1 g
T10235G 5 g
T102325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T10231G Supplier TCI America Supplier No. T10231G
Only null left

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00007748
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Melting Point 170°C
Quantity 1 g
Formula Weight 240.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,5,5′-Tetramethylbenzidine
RTECSNumber DV2300000
TSCA Yes