CAS RN 54827-17-7
3,3',5,5'-Tetramethylbenzidine, 99+%
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine, 98%
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, standard sensitivity
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, standard sensitivity, Thermo Scientific Chemicals
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity, Thermo Scientific Chemicals
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, high sensitivity, Thermo Scientific Chemicals
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine 98.0+%, TCI America™
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine 98.0+%, TCI America™
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine, MP Biomedicals™
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
Sigma Aldrich 3,3',5,5'-Tetramethylbenzidine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.