Learn More
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, standard sensitivity, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J60461AP
Description
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
Specifications
| 3,3',5,5'-Tetramethylbenzidine soln. | |
| 54827-17-7 | |
| C16H20N2 | |
| 500 mL | |
| 2808541 | |
| UAIUNKRWKOVEES-UHFFFAOYSA-N | |
| 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline | |
| 41206 | |
| Liquid |
| Ready-to-Use, precipitating, standard sensitivity | |
| Colorless to Yellow | |
| MFCD00007748 | |
| UN2810 | |
| 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble | |
| CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C | |
| 240.35 | |
| 240.35 |
Chemical Identifiers
| 54827-17-7 | |
| 240.35 | |
| UAIUNKRWKOVEES-UHFFFAOYSA-N | |
| 41206 | |
| CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C |
| C16H20N2 | |
| MFCD00007748 | |
| 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble | |
| 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline |
Safety and Handling
GHS H Statement
H370-H312-H332
Causes damage to organs.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P307+P311-P311-P312-P330-P363-P501c
H302+H312-H331-H336-H370
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 259-364-6
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only