missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, standard sensitivity, Thermo Scientific Chemicals

Catalog No. AAJ60461AU
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60461AU 1000 mL
AAJ60461AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60461AU Supplier Thermo Scientific Chemicals Supplier No. J60461AU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00007748
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Color Colorless to Yellow
Quantity 1000 mL
UN Number UN2810
Beilstein 2808541
Formula Weight 240.35
Physical Form Liquid
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, standard sensitivity
Hazard Category H302+H312-H331-H336-H370
Hazard Statement GHS H Statement
H370-H312-H332
Causes damage to organs.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P307+P311-P311-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 259-364-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only