missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, high sensitivity, Thermo Scientific Chemicals

Catalog No. aaj61325ap
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1000 mL
125 mL
500 mL
Catalog No. Quantity
AAJ61325AP 500 mL
AAJ61325EQE 125 mL
AAJ61325AU 1000 mL
3 options

Catalog No. AAJ61325AP

Supplier: Thermo Scientific Chemicals J61325AP

Only null left

3,3’,5,5’-Tetramethylbenzidine (TMB, CAS # 54827-17-7) is a blue chromogenic substance used in staining techniques such as immunohistochemistry.

TMB is a substrate for horseradish peroxidase. Recommended for ELISA (microwell) procedures,3,3?,5,5?-Tetramethylbenzidine has been used in a study to demonstrate that by measuring substrate absorption values off the absorption maximum, the measuring range of any immunoenzymometric assay can be extended significantly without jeopardizing assay characteristics. 3,3?,5,5?-Tetramethylbenzidine has also been used in a study to understand the formation of CuS concave superstructures with peroxidase-like activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• TMB is an aromatic amine that acts as a hydrogen donor in horseradish peroxidase-catalyzed reactions
• The transfer of hydrogen results in a blue stain associated with the target of interest
• This pH-dependent color change can be confirmed using spectrophotometric methods
• TMB is commonly used in immunohistochemistry; TMB is light-sensitive, and all applications should be performed in the dark

Application

• TMB is suitable for immunohistochemistry and immunocytochemistry applications including ELISA and cell and tissue staining

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00007748
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Color Yellow
Quantity 500 mL
UN Number UN2810
Beilstein 2808541
Solubility Information Insoluble in water. Soluble in organic solvents such as DMSO,96 % Ethanol,Acetone,Chloroform,Toluene.
Formula Weight 240.35
Physical Form Liquid
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, high sensitivity
Hazard Category H226-H302+H312-H331-H336-H370
Hazard Statement GHS H Statement
H370-H312-H332
Causes damage to organs.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 259-364-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only