missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine, MP Biomedicals™

Catalog No. p-197669
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 x 100 mg
Catalog No. Quantity
ICN980502 10 x 100 mg
1 options

Catalog No. ICN980502

Supplier: MP Biomedicals Inc 08980502

Only null left

Excellent for ELISA procedures

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Color Colorless
Linear Formula C16H20N2
Quantity 10 x 100 mg
Formula Weight 240.3
Physical Form Liquid
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine