missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine, 98%

Catalog No. AAA1386803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1386803 1 g
AAA1386806 5 g
AAA1386814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1386803 Supplier Thermo Scientific Chemicals Supplier No. A1386803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

3,3',5,5'-Tetramethylbenzidine, 98%, or TMB, CAS # 54827-17-7, is a colorimetric reagent used in laboratory-based substrate detection. | CAS: 54827-17-7 | C16H20N2 | 240.35 g/mol

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes; particularly useful in ELISA techniques, blue result can be read spectrophotometrically at 370 or 650 nm3,3',5,5'-Tetramethylbenzidine is used as a clinical reagent for testing blood. It is also a chromogenic substrate which is used in staining procedures in immunohistochemistry. Further, it is used as a visualizing reagent in enzyme-linked immunosorbent assays. In addition to this, it is used as a replacement for carcinogenic compounds such as benzidine and o-phenylenediamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 3,3',5,5'-Tetramethylbenzidine, also known as 4,4'-Diamino-3,3',5,5'-tetramethylbiphenyl or TMB, is a colorimetric substrate for detection of horseradish peroxidase-labeled probes
  • Oxidation of 3,3',5,5'-Tetramethylbenzidine results in a pale blue end product that can be detected at 370 or 655 nm. Sulfuric acid (H2SO4) is used to stop the oxidation reaction and stabilize the reading, resulting in a yellow color that can be read at 450 nm

Application

  • 3,3',5,5'-Tetramethylbenzidine is used as a chromogenic visualization agent in enzyme-linked immunosorbent assays (ELISA) or immunohistochemistry (IHC) techniques
  • 3,3',5,5'-Tetramethylbenzidine has been used to detect molecules in aqueous samples such as copper ion and hypochlorite

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00007748
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Melting Point 168°C to 171°C
Quantity 1 g
Beilstein 2808541
Sensitivity Light and moisture sensitive
Solubility Information Soluble in ethyl acetate,ethanol,acetone,chloroform,toluene and dimethyl sulfoxide. Insoluble in water.
Formula Weight 240.35
Percent Purity 98%
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine
Hazard Category H315-H319-H332-H335-H341
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
EINECSNumber 259-364-6
RTECSNumber DV2300000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only