missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',5,5'-Tetra-tert-butyl-2,2'-dihydroxybiphenyl 98.0+%, TCI America™
$130.48 - $451.10
Chemical Identifiers
| CAS | 6390-69-8 |
|---|---|
| Molecular Formula | C28H42O2 |
| Molecular Weight (g/mol) | 410.64 |
| MDL Number | MFCD00085287 |
| InChI Key | GDGDLBOVIAWEAD-UHFFFAOYSA-N |
| PubChem CID | 80808 |
| IUPAC Name | 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-2,2'-diol |
| SMILES | CC(C)(C)C1=CC(=C(O)C(=C1)C1=C(O)C(=CC(=C1)C(C)(C)C)C(C)(C)C)C(C)(C)C |
Chemical Identifiers
| 6390-69-8 | |
| 410.64 | |
| GDGDLBOVIAWEAD-UHFFFAOYSA-N | |
| 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-2,2'-diol |
| C28H42O2 | |
| MFCD00085287 | |
| 80808 | |
| CC(C)(C)C1=CC(=C(O)C(=C1)C1=C(O)C(=CC(=C1)C(C)(C)C)C(C)(C)C)C(C)(C)C |
Specifications
| 6390-69-8 | |
| White | |
| MFCD00085287 | |
| 3,3′C,5,5′C-Tetra-tert-butylbiphenyl-2,2′C-diol | |
| CC(C)(C)C1=CC(=C(O)C(=C1)C1=C(O)C(=CC(=C1)C(C)(C)C)C(C)(C)C)C(C)(C)C | |
| 410.64 | |
| 410.64 | |
| Crystalline Powder |
| 200°C | |
| C28H42O2 | |
| 1 g | |
| GDGDLBOVIAWEAD-UHFFFAOYSA-N | |
| 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-2,2'-diol | |
| 80808 | |
| ≥98.0% (GC) | |
| 3,3′,5,5′-Tetra-tert-butyl-2,2′-dihydroxybiphenyl |
Safety and Handling
TSCA : Yes