accessibility menu, dialog, popup

UserName

CH3H3CCH3OHH3CH3CCH3HOCH3H3CCH3CH3CH3H3C

CAS RN 6390-69-8

IUPAC Name: 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-2,2'-diol
Synonyms: 3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-2,2'-diol
Molecular Weight (g/mol): 410.64
Molecular Formula: C28H42O2
InChi Key: GDGDLBOVIAWEAD-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=C(O)C(=C1)C1=C(O)C(=CC(=C1)C(C)(C)C)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results