missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',5,5'-Tetra-tert-butyl-2,2'-dihydroxybiphenyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 200°C |
| Color | White |
| Quantity | 5 g |
| Synonym | 3,3′,5,5′-Tetra-tert-butylbiphenyl-2,2′-diol |
| Formula Weight | 410.64 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,3′,5,5′-Tetra-tert-butyl-2,2′-dihydroxybiphenyl |
Safety and Handling
| TSCA | Yes |