missing translation for 'onlineSavingsMsg'
Learn More

3,3,3',3'-Tetramethyl-5,5',6,6'-tetrapropoxy-1,1'-spirobiindane 98.0+%, TCI America™

Catalog No. T147725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T147725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T147725G Supplier TCI America Supplier No. T147725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89929-65-7
Molecular Formula C33H48O4
Molecular Weight (g/mol) 508.743
MDL Number MFCD00191503
InChI Key XFCRUFLAYQTRRX-UHFFFAOYSA-N
Synonym 3,3,3′C,3′C-Tetramethyl-5,5′C,6,6′C-tetrapropoxy-1,1′C-spirobisindane
PubChem CID 635699
IUPAC Name 1,1,1',1'-tetramethyl-5,5',6,6'-tetrapropoxy-3,3'-spirobi[2H-indene]
SMILES CCCOC1=C(C=C2C(=C1)C(CC23CC(C4=CC(=C(C=C34)OCCC)OCCC)(C)C)(C)C)OCCC

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 25 g
Formula Weight 508.74
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,3′,3′-Tetramethyl-5,5′,6,6′-tetrapropoxy-1,1′-spirobiindane
EINECSNumber (4)-1416
TSCA No