accessibility menu, dialog, popup

UserName

CH3OOH3COH3CH3CH3CCH3CH3OCH3

CAS RN 89929-65-7

IUPAC Name: 3,3,3',3'-tetramethyl-5,5',6,6'-tetrapropoxy-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]
Synonyms: 3,3,3',3'-tetramethyl-5,5',6,6'-tetrapropoxy-2H,2'H-1,1'-spirobi[indene]
Molecular Weight (g/mol): 508.74
Molecular Formula: C33H48O4
InChi Key: XFCRUFLAYQTRRX-UHFFFAOYSA-N
SMILES: CCCOC1=C(OCCC)C=C2C(=C1)C(C)(C)CC21CC(C)(C)C2=CC(OCCC)=C(OCCC)C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results