missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Nitroethenyl)pyridine 98.0+%, TCI America™

Catalog No. N06095G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06095G Supplier TCI America Supplier No. N06095G
Only null left

Chemical Identifiers

CAS 3156-52-3
Molecular Formula C7H6N2O2
Molecular Weight (g/mol) 150.137
MDL Number MFCD00191563
InChI Key HIXBXAXEKNYDHU-HWKANZROSA-N
Synonym 3-(2-Nitrovinyl)pyridine, 2-(3-Pyridyl)-1-nitroethylene
PubChem CID 5373109
IUPAC Name 3-[(E)-2-nitroethenyl]pyridine
SMILES C1=CC(=CN=C1)C=C[N+](=O)[O-]

Specifications

Melting Point 142°C
Quantity 5 g
UN Number 1325
Formula Weight 150.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Nitroethenyl)pyridine