CAS RN 3156-52-3
IUPAC Name:
3-(2-nitroethenyl)pyridine
Synonyms:
3-(2-nitroethenyl)pyridine
Molecular Weight (g/mol):
150.14
Molecular Formula:
C7H6N2O2
InChi Key:
HIXBXAXEKNYDHU-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C=CC1=CN=CC=C1
1
–
1
of
1
results
1
3-(2-Nitroethenyl)pyridine 98.0+%, TCI America™
CAS: 3156-52-3 Molecular Formula: C7H6N2O2 Molecular Weight (g/mol): 150.137 MDL Number: MFCD00191563 InChI Key: HIXBXAXEKNYDHU-HWKANZROSA-N Synonym: 3-(2-Nitrovinyl)pyridine, 2-(3-Pyridyl)-1-nitroethylene PubChem CID: 5373109 IUPAC Name: 3-[(E)-2-nitroethenyl]pyridine SMILES: C1=CC(=CN=C1)C=C[N+](=O)[O-]