missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Naphthyl)-L-alanine, 97%
CAS: 58438-03-2 | C13H13NO2 | 215.25 g/mol
$88.79 - $1230.86
Chemical Identifiers
| CAS | 58438-03-2 |
|---|---|
| Molecular Formula | C13H13NO2 |
| Molecular Weight (g/mol) | 215.25 |
| MDL Number | MFCD00066087 |
| InChI Key | JPZXHKDZASGCLU-LBPRGKRZSA-N |
| Synonym | 3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid |
| PubChem CID | 185915 |
| IUPAC Name | (2S)-2-amino-3-naphthalen-2-ylpropanoic acid |
| SMILES | N[C@@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6335403
|
Thermo Scientific Chemicals
H6335403 |
1 g |
Each for $88.79
|
|
|||||
|
AAH6335406
|
Thermo Scientific Chemicals
H6335406 |
5 g |
Each for $285.52
|
|
|||||
|
AAH6335414
|
Thermo Scientific Chemicals
H6335414 |
25 g |
Each for $1,230.86
|
|
|||||
Description
3-(2-Naphthyl)-L-alanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(2-Naphthyl)-L-alanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in 1mol/L NaOH.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
| 58438-03-2 | |
| 215.25 | |
| JPZXHKDZASGCLU-LBPRGKRZSA-N | |
| 185915 | |
| N[C@@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O |
| C13H13NO2 | |
| MFCD00066087 | |
| 3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid | |
| (2S)-2-amino-3-naphthalen-2-ylpropanoic acid |
Specifications
| 58438-03-2 | |
| MFCD00066087 | |
| 3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid | |
| JPZXHKDZASGCLU-LBPRGKRZSA-N | |
| (2S)-2-amino-3-naphthalen-2-ylpropanoic acid | |
| 215.25 | |
| 215.25 | |
| Solid |
| C13H13NO2 | |
| 2726272 | |
| Soluble in 1mol/L NaOH. | |
| N[C@@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O | |
| 1 g | |
| 185915 | |
| 97% | |
| 3-(2-Naphthyl)-L-alanine |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only