missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-L-alanine, 97%

CAS: 58438-03-2 | C13H13NO2 | 215.25 g/mol

Supplier:  Thermo Scientific Chemicals H6335406

 View more versions of this product

Catalog No. AAH6335406


Only null left

Description

Description

3-(2-Naphthyl)-L-alanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Naphthyl)-L-alanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1mol/L NaOH.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

3-(2-Naphthyl)-L-alanine
C13H13NO2
2726272
Soluble in 1mol/L NaOH.
N[C@@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O
5 g
185915
97%
58438-03-2
MFCD00066087
3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid
JPZXHKDZASGCLU-LBPRGKRZSA-N
(2S)-2-amino-3-naphthalen-2-ylpropanoic acid
215.25
215.25
Solid

Chemical Identifiers

58438-03-2
215.25
JPZXHKDZASGCLU-LBPRGKRZSA-N
185915
N[C@@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O
C13H13NO2
MFCD00066087
3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid
(2S)-2-amino-3-naphthalen-2-ylpropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only