missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-D-alanine, 95%

CAS: 76985-09-6 | C13H13NO2 | 215.25 g/mol

$141.73 - $1,838.45

Chemical Identifiers

CAS 76985-09-6
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD00038546
InChI Key JPZXHKDZASGCLU-GFCCVEGCSA-N
Synonym 3-2-naphthyl-d-alanine, h-d-2-nal-oh, r-2-amino-3-naphthalen-2-yl propanoic acid, r-2-amino-3-naphthalene-2-yl-propionic acid, d-2-naphthylalanine, d-3-2-naphthyl-alanine, unii-571v312ymy, 2-d-naphthylalanine, 2r-2-amino-3-2-naphthyl propanoic acid, 2r-2-amino-3-naphthalen-2-ylpropanoic acid
PubChem CID 2733661
IUPAC Name (2R)-2-amino-3-naphthalen-2-ylpropanoic acid
SMILES [NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH6353203
View Documents Promotion Details
N/A 1 g
Each for $141.73
Only null left
 
AAH6353206
View Documents Promotion Details
N/A 5 g
Each for $510.10
Only null left
 
AAH6353214
View Documents Promotion Details
N/A 25 g
Each for $1,838.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

76985-09-6
215.25
JPZXHKDZASGCLU-GFCCVEGCSA-N
2733661
[NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O
C13H13NO2
MFCD00038546
3-2-naphthyl-d-alanine, h-d-2-nal-oh, r-2-amino-3-naphthalen-2-yl propanoic acid, r-2-amino-3-naphthalene-2-yl-propionic acid, d-2-naphthylalanine, d-3-2-naphthyl-alanine, unii-571v312ymy, 2-d-naphthylalanine, 2r-2-amino-3-2-naphthyl propanoic acid, 2r-2-amino-3-naphthalen-2-ylpropanoic acid
(2R)-2-amino-3-naphthalen-2-ylpropanoic acid

Specifications

76985-09-6
C13H13NO2
4352384
JPZXHKDZASGCLU-GFCCVEGCSA-N
(2R)-2-amino-3-naphthalen-2-ylpropanoic acid
2733661
95%
3-(2-Naphthyl)-D-alanine
White
MFCD00038546
3-2-naphthyl-d-alanine, h-d-2-nal-oh, r-2-amino-3-naphthalen-2-yl propanoic acid, r-2-amino-3-naphthalene-2-yl-propionic acid, d-2-naphthylalanine, d-3-2-naphthyl-alanine, unii-571v312ymy, 2-d-naphthylalanine, 2r-2-amino-3-2-naphthyl propanoic acid, 2r-2-amino-3-naphthalen-2-ylpropanoic acid
[NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O
215.25
215.25
Solid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 616-416-6

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only