missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-D-alanine, 95%

CAS: 76985-09-6 | C13H13NO2 | 215.25 g/mol

Supplier:  Thermo Scientific Chemicals H6353206

Quantity 5 g

 View more versions of this product

Catalog No. AAH6353206


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

76985-09-6
215.25
JPZXHKDZASGCLU-GFCCVEGCSA-N
2733661
[NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O
C13H13NO2
MFCD00038546
3-2-naphthyl-d-alanine, h-d-2-nal-oh, r-2-amino-3-naphthalen-2-yl propanoic acid, r-2-amino-3-naphthalene-2-yl-propionic acid, d-2-naphthylalanine, d-3-2-naphthyl-alanine, unii-571v312ymy, 2-d-naphthylalanine, 2r-2-amino-3-2-naphthyl propanoic acid, 2r-2-amino-3-naphthalen-2-ylpropanoic acid
(2R)-2-amino-3-naphthalen-2-ylpropanoic acid

Specifications

3-(2-Naphthyl)-D-alanine
White
MFCD00038546
3-2-naphthyl-d-alanine, h-d-2-nal-oh, r-2-amino-3-naphthalen-2-yl propanoic acid, r-2-amino-3-naphthalene-2-yl-propionic acid, d-2-naphthylalanine, d-3-2-naphthyl-alanine, unii-571v312ymy, 2-d-naphthylalanine, 2r-2-amino-3-2-naphthyl propanoic acid, 2r-2-amino-3-naphthalen-2-ylpropanoic acid
[NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O
5 g
2733661
95%
76985-09-6
C13H13NO2
4352384
JPZXHKDZASGCLU-GFCCVEGCSA-N
(2R)-2-amino-3-naphthalen-2-ylpropanoic acid
215.25
215.25
Solid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 616-416-6

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions