missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyphenyl)propionic acid, 98+%

CAS: 6342-77-4 | C10H12O3 | 180.203 g/mol

Supplier:  Thermo Scientific Chemicals A1431406

 View more versions of this product

Catalog No. AAA1431406


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(2-Methoxyphenyl)propionic acid
85°C to 88°C
MFCD00002772
2097012
XSZSNLOPIWWFHS-UHFFFAOYSA-N
3-(2-methoxyphenyl)propanoic acid
80652
≥98%
6342-77-4
C10H12O3
5 g
3-2-methoxyphenyl propanoic acid, 3-2-methoxyphenyl propionic acid, 2-methoxyhydrocinnamic acid, benzenepropanoic acid, 2-methoxy, o-methoxyhydrocinnamic acid, 3-o-methoxyphenyl propionic acid, 2-methoxy-benzenepropanoic acid, beta-o-methoxyphenyl propionic acid, acmc-209nfi, maybridge1_000611
COC1=CC=CC=C1CCC(=O)O
180.203
180.2

Chemical Identifiers

6342-77-4
180.203
XSZSNLOPIWWFHS-UHFFFAOYSA-N
80652
COC1=CC=CC=C1CCC(=O)O
C10H12O3
MFCD00002772
3-2-methoxyphenyl propanoic acid, 3-2-methoxyphenyl propionic acid, 2-methoxyhydrocinnamic acid, benzenepropanoic acid, 2-methoxy, o-methoxyhydrocinnamic acid, 3-o-methoxyphenyl propionic acid, 2-methoxy-benzenepropanoic acid, beta-o-methoxyphenyl propionic acid, acmc-209nfi, maybridge1_000611
3-(2-methoxyphenyl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 228-738-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only