missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyphenyl)propionic acid, 98+%

Catalog No. p-7021820
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1431406 5 g
AAA1431414 25 g
AAA1431422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1431406 Supplier Thermo Scientific Chemicals Supplier No. A1431406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6342-77-4 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6342-77-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00002772
InChI Key XSZSNLOPIWWFHS-UHFFFAOYSA-N
Synonym 3-2-methoxyphenyl propanoic acid, 3-2-methoxyphenyl propionic acid, 2-methoxyhydrocinnamic acid, benzenepropanoic acid, 2-methoxy, o-methoxyhydrocinnamic acid, 3-o-methoxyphenyl propionic acid, 2-methoxy-benzenepropanoic acid, beta-o-methoxyphenyl propionic acid, acmc-209nfi, maybridge1_000611
PubChem CID 80652
IUPAC Name 3-(2-methoxyphenyl)propanoic acid
SMILES COC1=CC=CC=C1CCC(=O)O

Specifications

Melting Point 85°C to 88°C
Quantity 5 g
Beilstein 2097012
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material 3-(2-Methoxyphenyl)propionic acid
EINECSNumber 228-738-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only